missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyridoxal Hydrochloride 98.0+%, TCI America™
Specifications
Specifications
| Color | White-Yellow |
| Quantity | 5 g |
| Formula Weight | 203.62 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Grade | Research |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Pyridoxal Hydrochloride |
Safety and Handling
| EINECSNumber | (9)-1040 |
| RTECSNumber | UV1225000 |
| TSCA | Yes |