missing translation for 'onlineSavingsMsg'
Learn More

Pyridoxal hydrochloride, 99%

CAS: 65-22-5 | C8H9NO3·HCl | 203.62 g/mol

$77.61 - $6024.64

Chemical Identifiers

CAS 65-22-5
Molecular Formula C8H9NO3·HCl
Molecular Weight (g/mol) 203.62
MDL Number MFCD00012809
InChI Key FCHXJFJNDJXENQ-UHFFFAOYSA-N
Synonym pyridoxal hydrochloride, pyridoxal hcl, vitamin b6 hydrochloride, 3-hydroxy-5-hydroxymethyl-2-methylisonicotinaldehyde hydrochloride, unii-1416kf0qbc, 2-methyl-3-hydroxy-4-formyl-5-hydroxymethylpyridine hydrochloride, pl hcl, 3-hydroxy-5-hydroxymethyl-2-methylisonicotinaldehyde, hydrochloride, 3-hydroxy-5-hydroxymethyl-2-methyl-4-pyridinecarboxaldehyde hydrochloride, 3-hydroxy-5-hydroxymethyl-2-methylpyridine-4-carbaldehyde hydrochloride
PubChem CID 6171
IUPAC Name 3-hydroxy-5-(hydroxymethyl)-2-methylpyridine-4-carbaldehyde;hydrochloride
SMILES CC1=NC=C(C(=C1O)C=O)CO.Cl
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC352710010
SDS View Documents
Thermo Scientific Chemicals
352710010
1 g Glass bottle
Each for $77.61
Only null left
 
AC352710050
SDS View Documents
Thermo Scientific Chemicals
352710050
5 g Glass bottle
Each for $131.94
Only null left
 
AC352710250
SDS View Documents
Thermo Scientific Chemicals
352710250
25 g Glass bottle
Each for $449.01
Only null left
 
AC352715000
SDS View Documents
Thermo Scientific Chemicals
352715000
500 g Glass bottle
Each for $6,024.64
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

65-22-5
203.62
FCHXJFJNDJXENQ-UHFFFAOYSA-N
6171
CC1=NC=C(C(=C1O)C=O)CO.Cl
C8H9NO3·HCl
MFCD00012809
pyridoxal hydrochloride, pyridoxal hcl, vitamin b6 hydrochloride, 3-hydroxy-5-hydroxymethyl-2-methylisonicotinaldehyde hydrochloride, unii-1416kf0qbc, 2-methyl-3-hydroxy-4-formyl-5-hydroxymethylpyridine hydrochloride, pl hcl, 3-hydroxy-5-hydroxymethyl-2-methylisonicotinaldehyde, hydrochloride, 3-hydroxy-5-hydroxymethyl-2-methyl-4-pyridinecarboxaldehyde hydrochloride, 3-hydroxy-5-hydroxymethyl-2-methylpyridine-4-carbaldehyde hydrochloride
3-hydroxy-5-(hydroxymethyl)-2-methylpyridine-4-carbaldehyde;hydrochloride

Specifications

65-22-5
White
99%
C8H9NO3·HCl
1 g
pyridoxal hydrochloride, pyridoxal hcl, vitamin b6 hydrochloride, 3-hydroxy-5-hydroxymethyl-2-methylisonicotinaldehyde hydrochloride, unii-1416kf0qbc, 2-methyl-3-hydroxy-4-formyl-5-hydroxymethylpyridine hydrochloride, pl hcl, 3-hydroxy-5-hydroxymethyl-2-methylisonicotinaldehyde, hydrochloride, 3-hydroxy-5-hydroxymethyl-2-methyl-4-pyridinecarboxaldehyde hydrochloride, 3-hydroxy-5-hydroxymethyl-2-methylpyridine-4-carbaldehyde hydrochloride
FCHXJFJNDJXENQ-UHFFFAOYSA-N
3-hydroxy-5-(hydroxymethyl)-2-methylpyridine-4-carbaldehyde;hydrochloride
6171
99%
Pyridoxal hydrochloride
171.0°C to 175.0°C
Authentic
Glass bottle
MFCD00012809
15,8091
Solubility in water: 500g/L (20°C). Other solubilities: soluble in 95% ethanol: 1.7g/100 mL
CC1=NC=C(C(=C1O)C=O)CO.Cl
203.62
203.62
Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 200-602-5

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only