missing translation for 'onlineSavingsMsg'
Learn More

Pyridoxal hydrochloride, 99%

Catalog No. AAA1785506
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1785506 5 g
AAA1785514 25 g
AAA1785522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1785506 Supplier Thermo Scientific Chemicals Supplier No. A1785506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 65-22-5 | C8H10ClNO3 | 203.622 g/mol

Pyridoxal hydrochloride is used in labeling of amino acids for their detection. It finds application in neurotransmitters, sphingolipids and aminolevulinic acid. It is also used as a nutritional agent. It is also applied in research studies like biochemical, drugs and in media supplements.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyridoxal hydrochloride is used in labeling of amino acids for their detection. It finds application in neurotransmitters, sphingolipids and aminolevulinic acid. It is also used as a nutritional agent. It is also applied in research studies like biochemical, drugs and in media supplements.

Solubility
Soluble in water and ethanol.

Notes
Hygroscopic. Incompatible with strong oxidizing agents, strong acids, strong reducing agents and strong bases. Store in a cool place.

Chemical Identifiers

CAS 65-22-5
Molecular Formula C8H10ClNO3
Molecular Weight (g/mol) 203.622
MDL Number MFCD00012809
InChI Key FCHXJFJNDJXENQ-UHFFFAOYSA-N
Synonym pyridoxal hydrochloride, pyridoxal hcl, vitamin b6 hydrochloride, 3-hydroxy-5-hydroxymethyl-2-methylisonicotinaldehyde hydrochloride, unii-1416kf0qbc, 2-methyl-3-hydroxy-4-formyl-5-hydroxymethylpyridine hydrochloride, pl hcl, 3-hydroxy-5-hydroxymethyl-2-methylisonicotinaldehyde, hydrochloride, 3-hydroxy-5-hydroxymethyl-2-methyl-4-pyridinecarboxaldehyde hydrochloride, 3-hydroxy-5-hydroxymethyl-2-methylpyridine-4-carbaldehyde hydrochloride
PubChem CID 6171
IUPAC Name 3-hydroxy-5-(hydroxymethyl)-2-methylpyridine-4-carbaldehyde;hydrochloride
SMILES CC1=NC=C(C(=C1O)C=O)CO.Cl

Specifications

Melting Point 172°C to 174°C (decomposition)
Odor Odorless
Quantity 5 g
Beilstein 3656994
Sensitivity Hygroscopic
Merck Index 14,7978
Solubility Information Soluble in water and ethanol.
Formula Weight 203.63
Percent Purity 99%
Chemical Name or Material Pyridoxal hydrochloride
EINECSNumber 200-602-5
RTECSNumber UV1225000
TSCA Yes
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only