missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyridoxal 5-Phosphate Monohydrate 98.0+%, TCI America™
Supplier: TCI America C03775G
Specifications
| Pyridoxal 5-Phosphate Monohydrate | |
| 143°C | |
| C8H10NO6P | |
| 5 g | |
| NGVDGCNFYWLIFO-UHFFFAOYSA-N | |
| [(4-formyl-5-hydroxy-6-methylpyridin-3-yl)methoxy]phosphonic acid | |
| 38882 | |
| ≥98.0% (HPLC) |
| 41468-25-1 | |
| Yellow | |
| MFCD00006333 | |
| 4-formyl-5-hydroxy-6-methylpyridin-3-yl methyl dihydrogen phosphate hydrate, pyridoxal phosphate hydrate, pyridoxal 5-phosphate monohydrate, phosphopyridoxal monohydrate, pyridoxyl phosphate monohydrate, unii-5v5ioj8338, pyridoxal-5-phosphate, monohydrate, pyridoxal-5'-phosphate monohydrate, pyridoxal-5-phosphate monohydrate, pyridoxal 5'-phosphate monohydrate | |
| CC1=NC=C(COP(O)(O)=O)C(C=O)=C1O | |
| 247.14 | |
| 247.14 | |
| Crystalline Powder |
Chemical Identifiers
| 41468-25-1 | |
| 247.14 | |
| NGVDGCNFYWLIFO-UHFFFAOYSA-N | |
| 38882 | |
| CC1=NC=C(COP(O)(O)=O)C(C=O)=C1O |
| C8H10NO6P | |
| MFCD00006333 | |
| 4-formyl-5-hydroxy-6-methylpyridin-3-yl methyl dihydrogen phosphate hydrate, pyridoxal phosphate hydrate, pyridoxal 5-phosphate monohydrate, phosphopyridoxal monohydrate, pyridoxyl phosphate monohydrate, unii-5v5ioj8338, pyridoxal-5-phosphate, monohydrate, pyridoxal-5'-phosphate monohydrate, pyridoxal-5-phosphate monohydrate, pyridoxal 5'-phosphate monohydrate | |
| [(4-formyl-5-hydroxy-6-methylpyridin-3-yl)methoxy]phosphonic acid |
Safety and Handling
EINECSNumber : (9)-1512
RTECSNumber : UV1208000
TSCA : Yes