accessibility menu, dialog, popup

UserName

CH3NOPHOOHOOOH

CAS RN 41468-25-1

IUPAC Name: [(4-formyl-5-hydroxy-6-methylpyridin-3-yl)methoxy]phosphonic acid
Synonyms: pyridoxal phosphate|pyridoxal-5'-phosphate|pyridoxal 5 phosphate|pyridoxal 5-phosphate|pyridoxal-5-phosphate|pyridoxal 5'-phosphate|pyridoxal-P|pyridoxal P|phosphate, pyridoxal|pal-P
Molecular Weight (g/mol): 247.14
Molecular Formula: C8H10NO6P
InChi Key: NGVDGCNFYWLIFO-UHFFFAOYSA-N
SMILES: CC1=NC=C(COP(O)(O)=O)C(C=O)=C1O
Molecular Weight (g/mol) 
  • (10)
  • (4)
Percent Purity 
  • (3)
  • (3)
  • (4)
  • (3)
Quantity 
  • (3)
  • (3)
  • (4)
Physical Form 
  • (6)
  • (1)
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (4)
  • (4)
  • (1)
  • (3)
  • (6)
  • (10)
  • (4)
  • (3)
  • (3)
  • (4)
  • (3)
  • (3)
  • (3)
  • (4)
  • (6)
  • (1)
  • (4)