missing translation for 'onlineSavingsMsg'
Learn More

Pyridoxal 5-Phosphate Monohydrate 98.0+%, TCI America™

Catalog No. C03771G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C03771G 1 g
C03775G 5 g
C037725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C03771G Supplier TCI America Supplier No. C03771G

Chemical Identifiers

CAS 41468-25-1
Molecular Formula C8H10NO6P
Molecular Weight (g/mol) 247.14
MDL Number MFCD00006333
InChI Key NGVDGCNFYWLIFO-UHFFFAOYSA-N
Synonym 4-formyl-5-hydroxy-6-methylpyridin-3-yl methyl dihydrogen phosphate hydrate, pyridoxal phosphate hydrate, pyridoxal 5-phosphate monohydrate, phosphopyridoxal monohydrate, pyridoxyl phosphate monohydrate, unii-5v5ioj8338, pyridoxal-5-phosphate, monohydrate, pyridoxal-5'-phosphate monohydrate, pyridoxal-5-phosphate monohydrate, pyridoxal 5'-phosphate monohydrate
PubChem CID 38882
IUPAC Name [(4-formyl-5-hydroxy-6-methylpyridin-3-yl)methoxy]phosphonic acid
SMILES CC1=NC=C(COP(O)(O)=O)C(C=O)=C1O

Specifications

Melting Point 143°C
Color Yellow
Quantity 1 g
Formula Weight 247.14
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Pyridoxal 5-Phosphate Monohydrate
EINECSNumber (9)-1512
RTECSNumber UV1208000
TSCA Yes