missing translation for 'onlineSavingsMsg'
Learn More

Medisca Pyridoxal-5-Phosphate (Monohydrate) (Dietary Supplement Grade)

Catalog No. 14186243
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
500 g
1 kg
5 kg
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14186243 500 g
14186116 1 kg
14186164 1 g
14186098 100 g
14185477 25 g
14185594 5 g
14185593 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 14186243 Supplier Medisca Supplier No. 155608*

Pyridoxal-5-Phosphate (Monohydrate) (Dietary Supplement Grade) is a slightly yellow-white to pale yellow, odorless crystalline powder.

Pyridoxal-5-Phosphate (Monohydrate) is a slightly yellow-white to pale yellow, odorless crystalline powder.

Dietary Supplement Grade refers to a nutritional supplement that is manufactured by an FDA-registered dietary supplement manufacturer or manufacturer with other certifications and registrations, based on applicable laws and regulations. Qualified manufacturers follow their applicable standards of practice, which are less stringent than those for FDA-registered drug manufacturers. The customer is responsible for ensuring suitability of the product for the intended use and acknowledges that further testing may be required.

Specifications

Chemical Name or Material Pyridoxal-5-Phosphate (Monohydrate) (Dietary Supplement Grade)
Packaging Jar, 1500mL, White, HDPE, Round, 89-400, TE Pano Neck
CAS 41468-25-1
Type Drug Standard
Molecular Weight (g/mol) 265.1568
Quantity 500 g
Percent Purity 99 to 101 % (Pyridoxal-5-phosphate)
Grade Ungraded