missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Medisca Pyridoxal-5-Phosphate (Monohydrate) (Dietary Supplement Grade)
Description
Pyridoxal-5-Phosphate (Monohydrate) is a slightly yellow-white to pale yellow, odorless crystalline powder.
Dietary Supplement Grade refers to a nutritional supplement that is manufactured by an FDA-registered dietary supplement manufacturer or manufacturer with other certifications and registrations, based on applicable laws and regulations. Qualified manufacturers follow their applicable standards of practice, which are less stringent than those for FDA-registered drug manufacturers. The customer is responsible for ensuring suitability of the product for the intended use and acknowledges that further testing may be required.
Specifications
Specifications
| Chemical Name or Material | Pyridoxal-5-Phosphate (Monohydrate) (Dietary Supplement Grade) |
| Packaging | Jar, 2.8L, White, HDPE, Round, 110-400, TE Pano Neck |
| CAS | 41468-25-1 |
| Type | Drug Standard |
| Molecular Weight (g/mol) | 265.1568 |
| Quantity | 1 kg |
| Percent Purity | 99 to 101 % (Pyridoxal-5-phosphate) |
| Grade | Ungraded |