missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyridoxal-5-Phosphate Monohydrate, Approx. 98%, MP Biomedicals™
$503.76 - $503.76
Chemical Identifiers
| CAS | 41468-25-1 |
|---|---|
| Molecular Formula | C8H10NO6P |
| Molecular Weight (g/mol) | 247.14 |
| MDL Number | MFCD00006333 |
| InChI Key | NGVDGCNFYWLIFO-UHFFFAOYSA-N |
| Synonym | 4-formyl-5-hydroxy-6-methylpyridin-3-yl methyl dihydrogen phosphate hydrate, pyridoxal phosphate hydrate, pyridoxal 5-phosphate monohydrate, phosphopyridoxal monohydrate, pyridoxyl phosphate monohydrate, unii-5v5ioj8338, pyridoxal-5-phosphate, monohydrate, pyridoxal-5'-phosphate monohydrate, pyridoxal-5-phosphate monohydrate, pyridoxal 5'-phosphate monohydrate |
| PubChem CID | 38882 |
| IUPAC Name | [(4-formyl-5-hydroxy-6-methylpyridin-3-yl)methoxy]phosphonic acid |
| SMILES | CC1=NC=C(COP(O)(O)=O)C(C=O)=C1O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN10276905
|
MP Biomedicals Inc
0210276905 |
5 g |
Each for $503.76
|
|
|||||
Description
CoenzymeChemical Identifiers
| 41468-25-1 | |
| 247.14 | |
| NGVDGCNFYWLIFO-UHFFFAOYSA-N | |
| 38882 | |
| CC1=NC=C(COP(O)(O)=O)C(C=O)=C1O |
| C8H10NO6P | |
| MFCD00006333 | |
| 4-formyl-5-hydroxy-6-methylpyridin-3-yl methyl dihydrogen phosphate hydrate, pyridoxal phosphate hydrate, pyridoxal 5-phosphate monohydrate, phosphopyridoxal monohydrate, pyridoxyl phosphate monohydrate, unii-5v5ioj8338, pyridoxal-5-phosphate, monohydrate, pyridoxal-5'-phosphate monohydrate, pyridoxal-5-phosphate monohydrate, pyridoxal 5'-phosphate monohydrate | |
| [(4-formyl-5-hydroxy-6-methylpyridin-3-yl)methoxy]phosphonic acid |
Specifications
| 41468-25-1 | |
| appox. 98% | |
| MFCD00006333 | |
| 4-formyl-5-hydroxy-6-methylpyridin-3-yl methyl dihydrogen phosphate hydrate, pyridoxal phosphate hydrate, pyridoxal 5-phosphate monohydrate, phosphopyridoxal monohydrate, pyridoxyl phosphate monohydrate, unii-5v5ioj8338, pyridoxal-5-phosphate, monohydrate, pyridoxal-5'-phosphate monohydrate, pyridoxal-5-phosphate monohydrate, pyridoxal 5'-phosphate monohydrate | |
| CC1=NC=C(COP(O)(O)=O)C(C=O)=C1O | |
| 247.14 | |
| 265.1 | |
| Pyridoxal-5-Phosphate Monohydrate |
| White | |
| C8H10NO6P | |
| 5 g | |
| NGVDGCNFYWLIFO-UHFFFAOYSA-N | |
| [(4-formyl-5-hydroxy-6-methylpyridin-3-yl)methoxy]phosphonic acid | |
| 38882 | |
| Solid |
Safety and Handling
Recommended Storage : Store at -0°C.