missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyridinium Trifluoromethanesulfonate 98.0+%, TCI America™
$98.18 - $311.89
Chemical Identifiers
| CAS | 52193-54-1 |
|---|---|
| Molecular Formula | C6H6F3NO3S |
| Synonym | pyridinium trifluoromethanesulfonate, pyridinium triflate, pyridin-1-ium trifluoromethanesulfonate, pyridium triflate, acmc-209ky5, pyridinium trifluoromethanesulphonate, pyridin-1-ium; trifluoromethanesulfonate |
Chemical Identifiers
| 52193-54-1 | |
| C6H6F3NO3S | |
| pyridinium trifluoromethanesulfonate, pyridinium triflate, pyridin-1-ium trifluoromethanesulfonate, pyridium triflate, acmc-209ky5, pyridinium trifluoromethanesulphonate, pyridin-1-ium; trifluoromethanesulfonate |
Specifications
| 52193-54-1 | |
| White | |
| 5 g | |
| 229.17 | |
| Crystalline Powder |
| 224°C | |
| C6H6F3NO3S | |
| pyridinium trifluoromethanesulfonate, pyridinium triflate, pyridin-1-ium trifluoromethanesulfonate, pyridium triflate, acmc-209ky5, pyridinium trifluoromethanesulphonate, pyridin-1-ium; trifluoromethanesulfonate | |
| ≥98.0% (HPLC,T) | |
| Pyridinium Trifluoromethanesulfonate |
Safety and Handling
TSCA : No