missing translation for 'onlineSavingsMsg'
Learn More

Pyridinium p-Toluenesulfonate 98.0+%, TCI America™

Catalog No. P094225G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P094225G 25 g
P0942100G 100 g
P0942500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P094225G Supplier TCI America Supplier No. P094225G

Chemical Identifiers

CAS 24057-28-1
Molecular Formula C12H13NO3S
MDL Number MFCD00013108
Synonym pyridinium p-toluenesulfonate, ppts, pyridin-1-ium 4-methylbenzenesulfonate, pyridinium 4-toluenesulfonate, pyridinium tosylate, pyridinium 4-methylbenzenesulfonate, pyridinium p-toluene sulfonate, pyridinium p-toluene sulphonate, pyridinium para-toluenesulfonate, 4-toluenesulfonic acid pyridine salt

Specifications

Melting Point 120°C
Color White-Yellow
Quantity 25 g
Formula Weight 251.30
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Pyridinium p-Toluenesulfonate