Learn More
Pyridinium p-toluenesulfonate, 98+%
CAS: 24057-28-1 | C12H13NO3S
Supplier: Thermo Scientific Chemicals A1570822
Description
Pyridinium p-Toluenesulfate is used in the synthesis of a subdomain of the cytochrome P450 BM3 enzyme, for use in screening systems for drug candidates. Efficient catalyst for preparing tetrahydropyranyl ethers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPyridinium p-Toluenesulfate is used in the synthesis of a subdomain of the cytochrome P450 BM3 enzyme, for use in screening systems for drug candidates. Efficient catalyst for preparing tetrahydropyranyl ethers.
Solubility
Soluble in water (partly), ethanol, acetone, dichloromethane, chloroform, methanol and dimethyl sulfoxide.
Notes
Moisture Sensitive. Store in a tightly closed container. Store in a cool, dry, well-ventilated. Store protected from moisture.
Specifications
| Pyridinium p-toluenesulfonate | |
| 116°C to 120°C | |
| MFCD00013108 | |
| 3764305 | |
| pyridinium p-toluenesulfonate, ppts, pyridin-1-ium 4-methylbenzenesulfonate, pyridinium 4-toluenesulfonate, pyridinium tosylate, pyridinium 4-methylbenzenesulfonate, pyridinium p-toluene sulfonate, pyridinium p-toluene sulphonate, pyridinium para-toluenesulfonate, 4-toluenesulfonic acid pyridine salt | |
| 251.31 |
| 24057-28-1 | |
| C12H13NO3S | |
| 100 g | |
| Moisture sensitive | |
| Soluble in water (partly),ethanol,acetone,dichloromethane,chloroform,methanol and dimethyl sulfoxide. | |
| ≥98% |
Chemical Identifiers
| 24057-28-1 | |
| C12H13NO3S | |
| MFCD00013108 | |
| pyridinium p-toluenesulfonate, ppts, pyridin-1-ium 4-methylbenzenesulfonate, pyridinium 4-toluenesulfonate, pyridinium tosylate, pyridinium 4-methylbenzenesulfonate, pyridinium p-toluene sulfonate, pyridinium p-toluene sulphonate, pyridinium para-toluenesulfonate, 4-toluenesulfonic acid pyridine salt |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 246-002-7
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only