missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyridinium Bromide Perbromide 85.0+%, TCI America™
Specifications
Specifications
| Melting Point | 136°C |
| Quantity | 25 g |
| UN Number | 1759 |
| Formula Weight | 319.82 |
| Percent Purity | ≥85.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Pyridinium Bromide Perbromide |
Safety and Handling
| TSCA | Yes |