missing translation for 'onlineSavingsMsg'
Learn More

Pyridinium Bromide Perbromide 85.0+%, TCI America™

Catalog No. P082525G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P082525G 25 g
P0825100G 100 g
P0825500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P082525G Supplier TCI America Supplier No. P082525G
Only null left

Chemical Identifiers

CAS 39416-48-3
Molecular Formula C5H6Br3N
MDL Number MFCD00013223
Synonym Pyridine Hydrobromide Perbromide, Pyridinium Tribromide

Specifications

Melting Point 136°C
Quantity 25 g
UN Number 1759
Formula Weight 319.82
Percent Purity ≥85.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Pyridinium Bromide Perbromide