missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyridine - Sulfur Trioxide Complex 95.0+%, TCI America™
$1263.83 - $1263.83
Chemical Identifiers
| CAS | 26412-87-3 |
|---|---|
| Molecular Formula | C5H5NO3S |
| Molecular Weight (g/mol) | 159.16 |
| MDL Number | MFCD00012437 |
| InChI Key | UDYFLDICVHJSOY-UHFFFAOYSA-N |
| Synonym | pyridine sulfur trioxide, sulfur trioxide pyridine complex, pyridine; sulfur trioxide, pyridine sulfotrioxide, pyridine compound with sulfur trioxide 1:1, sulfur trioxide, compd. with pyridine, pyridine-sulfur trioxide complex, active sulfure trioxide, sulfur trioxide-pyridine, pyridine-sulfur trioxide |
| PubChem CID | 168533 |
| IUPAC Name | pyridine; sulfonylideneoxidane |
| SMILES | O=S(=O)=O.C1=CC=NC=C1 |
Chemical Identifiers
| 26412-87-3 | |
| 159.16 | |
| UDYFLDICVHJSOY-UHFFFAOYSA-N | |
| 168533 | |
| O=S(=O)=O.C1=CC=NC=C1 |
| C5H5NO3S | |
| MFCD00012437 | |
| pyridine sulfur trioxide, sulfur trioxide pyridine complex, pyridine; sulfur trioxide, pyridine sulfotrioxide, pyridine compound with sulfur trioxide 1:1, sulfur trioxide, compd. with pyridine, pyridine-sulfur trioxide complex, active sulfure trioxide, sulfur trioxide-pyridine, pyridine-sulfur trioxide | |
| pyridine; sulfonylideneoxidane |
Specifications
| 26412-87-3 | |
| C5H5NO3S | |
| 500 g | |
| pyridine sulfur trioxide, sulfur trioxide pyridine complex, pyridine; sulfur trioxide, pyridine sulfotrioxide, pyridine compound with sulfur trioxide 1:1, sulfur trioxide, compd. with pyridine, pyridine-sulfur trioxide complex, active sulfure trioxide, sulfur trioxide-pyridine, pyridine-sulfur trioxide | |
| O=S(=O)=O.C1=CC=NC=C1 | |
| 159.16 | |
| 159.16 | |
| Crystalline Powder |
| White-Yellow | |
| MFCD00012437 | |
| 3261 | |
| UDYFLDICVHJSOY-UHFFFAOYSA-N | |
| pyridine; sulfonylideneoxidane | |
| 168533 | |
| ≥95.0% (T) | |
| Pyridine - Sulfur Trioxide Complex |
Safety and Handling
TSCA : Yes