accessibility menu, dialog, popup

UserName

CAS RN 26412-87-3

IUPAC Name: pyridine; sulfonylideneoxidane
Synonyms: sulfur trioxide pyridine complex
Molecular Weight (g/mol): 159.16
Molecular Formula: C5H5NO3S
InChi Key: UDYFLDICVHJSOY-UHFFFAOYSA-N
SMILES: O=S(=O)=O.C1=CC=NC=C1
Percent Purity 
  • (1)
  • (4)
  • (4)
Physical Form 
  • (1)
  • (8)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (8)
  • (1)
  • (8)
  • (1)
  • (4)
  • (4)
  • (1)
  • (8)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)