missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyridine Iodine Monochloride 98.0+%, TCI America™
Supplier: TCI America P20861G
Specifications
| Pyridine Iodine Monochloride | |
| 135°C | |
| C5H5ClIN | |
| 1 g | |
| SYWWBJKPSYWUBN-UHFFFAOYSA-N | |
| iodochlorane; pyridine | |
| 10220417 | |
| ≥98.0% (T) |
| 6443-90-9 | |
| White-Yellow | |
| MFCD06410987 | |
| Pyridine Iodochloride | |
| ClI.C1=CC=NC=C1 | |
| 241.46 | |
| 241.46 | |
| Crystalline Powder |
Chemical Identifiers
| 6443-90-9 | |
| 241.46 | |
| SYWWBJKPSYWUBN-UHFFFAOYSA-N | |
| 10220417 | |
| ClI.C1=CC=NC=C1 |
| C5H5ClIN | |
| MFCD06410987 | |
| Pyridine Iodochloride | |
| iodochlorane; pyridine |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator