missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyridine Iodine Monochloride 98.0+%, TCI America™
$277.75 - $1243.32
Chemical Identifiers
| CAS | 6443-90-9 |
|---|---|
| Molecular Formula | C5H5ClIN |
| Molecular Weight (g/mol) | 241.46 |
| MDL Number | MFCD06410987 |
| InChI Key | SYWWBJKPSYWUBN-UHFFFAOYSA-N |
| Synonym | Pyridine Iodochloride |
| PubChem CID | 10220417 |
| IUPAC Name | iodochlorane; pyridine |
| SMILES | ClI.C1=CC=NC=C1 |
Chemical Identifiers
| 6443-90-9 | |
| 241.46 | |
| SYWWBJKPSYWUBN-UHFFFAOYSA-N | |
| 10220417 | |
| ClI.C1=CC=NC=C1 |
| C5H5ClIN | |
| MFCD06410987 | |
| Pyridine Iodochloride | |
| iodochlorane; pyridine |
Specifications
| 6443-90-9 | |
| White-Yellow | |
| MFCD06410987 | |
| Pyridine Iodochloride | |
| ClI.C1=CC=NC=C1 | |
| 241.46 | |
| 241.46 | |
| Crystalline Powder |
| 135°C | |
| C5H5ClIN | |
| 1 g | |
| SYWWBJKPSYWUBN-UHFFFAOYSA-N | |
| iodochlorane; pyridine | |
| 10220417 | |
| ≥98.0% (T) | |
| Pyridine Iodine Monochloride |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator