missing translation for 'onlineSavingsMsg'
Learn More

Pyridine hydrochloride, 98%

CAS: 628-13-7 | C5H6ClN | 115.56 g/mol

Supplier:  Thermo Scientific Chemicals A108710B

 View more versions of this product

Catalog No. AAA108710B


Only null left
Explore available promotions

Description

Description

Pyridine hydrochloride is a classical reagent for cleavage of aryl methyl ethers at 200-220o. It is also used in the demethylation of 4,5-dimethyl-7-methoxy-1-tetralone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyridine hydrochloride is a classical reagent for cleavage of aryl methyl ethers at 200-220o. It is also used in the demethylation of 4,5-dimethyl-7-methoxy-1-tetralone.

Solubility
Soluble in water (85 g/100 ml) and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is hygroscopic in nature.
Specifications

Specifications

Pyridine hydrochloride
∼145°C
C5H6ClN
1000 g
Hygroscopic
Soluble in water (85g/100 ml) and methanol.
C1=CC=NC=C1.Cl
115.56
115.56
628-13-7
222°C to 224°C
MFCD00012802
3615340
pyridine hydrochloride, pyridine, hydrochloride, pyridinehydrochloride, pyridine hcl, pyridine, hydrochloride 1:1, pyridine-hcl, pyridine.hcl, pyridiniumchloride, pyridine hcl salt, pyrdine hydrochloride
AOJFQRQNPXYVLM-UHFFFAOYSA-N
pyridine;hydrochloride
10176127
98%

Chemical Identifiers

628-13-7
115.56
AOJFQRQNPXYVLM-UHFFFAOYSA-N
10176127
C1=CC=NC=C1.Cl
C5H6ClN
MFCD00012802
pyridine hydrochloride, pyridine, hydrochloride, pyridinehydrochloride, pyridine hcl, pyridine, hydrochloride 1:1, pyridine-hcl, pyridine.hcl, pyridiniumchloride, pyridine hcl salt, pyrdine hydrochloride
pyridine;hydrochloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

EINECSNumber : 211-027-4

RTECSNumber : UT4375000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only