missing translation for 'onlineSavingsMsg'
Learn More

Pyridine hydrochloride, 98%

Catalog No. p-7081061
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1087118 50 g
AAA1087130 250 g
AAA108710B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1087118 Supplier Thermo Scientific Chemicals Supplier No. A1087118

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 628-13-7 | C5H6ClN | 115.56 g/mol

Pyridine hydrochloride is a classical reagent for cleavage of aryl methyl ethers at 200-220o. It is also used in the demethylation of 4,5-dimethyl-7-methoxy-1-tetralone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyridine hydrochloride is a classical reagent for cleavage of aryl methyl ethers at 200-220o. It is also used in the demethylation of 4,5-dimethyl-7-methoxy-1-tetralone.

Solubility
Soluble in water (85 g/100 ml) and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is hygroscopic in nature.

Chemical Identifiers

CAS 628-13-7
Molecular Formula C5H6ClN
Molecular Weight (g/mol) 115.56
MDL Number MFCD00012802
InChI Key AOJFQRQNPXYVLM-UHFFFAOYSA-N
Synonym pyridine hydrochloride, pyridine, hydrochloride, pyridinehydrochloride, pyridine hcl, pyridine, hydrochloride 1:1, pyridine-hcl, pyridine.hcl, pyridiniumchloride, pyridine hcl salt, pyrdine hydrochloride
PubChem CID 10176127
IUPAC Name pyridine;hydrochloride
SMILES C1=CC=NC=C1.Cl

Specifications

Melting Point ∼145°C
Boiling Point 222°C to 224°C
Quantity 50 g
Beilstein 3615340
Sensitivity Hygroscopic
Solubility Information Soluble in water (85g/100 ml) and methanol.
Formula Weight 115.56
Percent Purity 98%
Chemical Name or Material Pyridine hydrochloride
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 211-027-4
RTECSNumber UT4375000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only