missing translation for 'onlineSavingsMsg'
Learn More

Pyridine hydrobromide perbromide, tech. 90%

CAS: 39416-48-3 | C5H5N·HBr3

$85.45 - $646.10

Chemical Identifiers

CAS 39416-48-3
Molecular Formula C5H5N·HBr3
MDL Number MFCD00013223
Synonym Pyridinium bromide perbromide; Pyridinium tribromide
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1568414
View Documents
Thermo Scientific Chemicals
A1568414
25 g
Each for $85.45
Only null left
 
AAA1568422
View Documents
Thermo Scientific Chemicals
A1568422
100 g
Each for $199.48
Only null left
 
AAA1568436
View Documents
Thermo Scientific Chemicals
A1568436
500 g
Each for $646.10
Only null left
 
Description

Description

Pyridine hydrobromide perbromide is used as a brominating reagent in alfa-bromination and alfa-thiocyanation of ketones, phenols, unsaturated and aromatic ethers. It is used as a raw material in the preparation of beta-adrenergic blocking agents. Furthermore, it is used as an analytical reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyridine hydrobromide perbromide is used as a brominating reagent in alfa-bromination and alfa-thiocyanation of ketones, phenols, unsaturated and aromatic ethers. It is used as a raw material in the preparation of beta-adrenergic blocking agents. Furthermore, it is used as an analytical reagent.

Solubility
Soluble in methanol, acetic acid, ethanol, n-butanol and terahydrofuran. Insoluble in water, carbon tetrachloride, ethyl bromide, benzene, toluene, ligroin, petroleum ether.

Notes
Store in cool place. Incompatible with strong oxidizing agents and strong bases.
Specifications

Chemical Identifiers

39416-48-3
C5H5N·HBr3
MFCD00013223
Pyridinium bromide perbromide; Pyridinium tribromide

Specifications

39416-48-3
C5H5N·HBr3
25 g
3690144
Pyridinium bromide perbromide; Pyridinium tribromide
319.82
Technical
128°C to 136°C
MFCD00013223
UN3261
14,7973
Soluble in methanol,acetic acid,ethanol,n-butanol and terahydrofuran. Insoluble in water,carbon tetrachloride,ethyl bromide,benzene,toluene,ligroin,petroleum ether.
90%
Pyridine hydrobromide perbromide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 254-446-8

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only