missing translation for 'onlineSavingsMsg'
Learn More

Pyridine hydrobromide perbromide, tech. 90%

Catalog No. AAA1568436
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1568436 500 g
AAA1568414 25 g
AAA1568422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1568436 Supplier Thermo Scientific Chemicals Supplier No. A1568436

CAS: 39416-48-3 | C5H5N·HBr3

Pyridine hydrobromide perbromide is used as a brominating reagent in alfa-bromination and alfa-thiocyanation of ketones, phenols, unsaturated and aromatic ethers. It is used as a raw material in the preparation of beta-adrenergic blocking agents. Furthermore, it is used as an analytical reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyridine hydrobromide perbromide is used as a brominating reagent in alfa-bromination and alfa-thiocyanation of ketones, phenols, unsaturated and aromatic ethers. It is used as a raw material in the preparation of beta-adrenergic blocking agents. Furthermore, it is used as an analytical reagent.

Solubility
Soluble in methanol, acetic acid, ethanol, n-butanol and terahydrofuran. Insoluble in water, carbon tetrachloride, ethyl bromide, benzene, toluene, ligroin, petroleum ether.

Notes
Store in cool place. Incompatible with strong oxidizing agents and strong bases.

Chemical Identifiers

CAS 39416-48-3
Molecular Formula C5H5N·HBr3
MDL Number MFCD00013223
Synonym Pyridinium bromide perbromide; Pyridinium tribromide

Specifications

Melting Point 128°C to 136°C
Quantity 500 g
UN Number UN3261
Beilstein 3690144
Merck Index 14,7973
Solubility Information Soluble in methanol,acetic acid,ethanol,n-butanol and terahydrofuran. Insoluble in water,carbon tetrachloride,ethyl bromide,benzene,toluene,ligroin,petroleum ether.
Formula Weight 319.82
Percent Purity 90%
Grade Technical
Chemical Name or Material Pyridine hydrobromide perbromide
Hazard Category H302-H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 254-446-8
TSCA Yes

RUO – Research Use Only