missing translation for 'onlineSavingsMsg'
Learn More

Pyridine, CHROMASOLV™ Plus, for HPLC, ≥99.9%, Honeywell Riedel-de Haën™

CHROMASOLV™ Plus, for HPLC,≥99.9%

$667.08 - $667.08

Chemical Identifiers

CAS 110-86-1
Molecular Formula C5H5N
Molecular Weight (g/mol) 79.102
MDL Number MFCD00011732
InChI Key JUJWROOIHBZHMG-UHFFFAOYSA-N
Synonym azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717
PubChem CID 1049
ChEBI CHEBI:16227
IUPAC Name pyridine
SMILES C1=CC=NC=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
6001314
View Documents
Honeywell Chemicals
2704071L
1 L Glass Bottle N/A
Only null left
 
Description

Description

  • For HPLC, spectrophotometry, and environmental analysis
  • Tested for HPLC-suitability via gradient analysis and UV-VIS spectroscopy
  • Low UV impurity profile
  • Low impurity gradient LC baseline
  • Very low levels of non-volatile impurities
Specifications

Chemical Identifiers

110-86-1
79.102
JUJWROOIHBZHMG-UHFFFAOYSA-N
1049
pyridine
C5H5N
MFCD00011732
azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717
CHEBI:16227
C1=CC=NC=C1

Specifications

-42°C (literature)
max. 0.01 at 400 nm, max. 0.01 at 350 nm, max. 0.02 at 335 nm, max. 0.15 at 315 nm, max. 1.0 at 305 nm
0.978 g/mL at 25 °C(lit.)
1 L
Glass Bottle
C5H5N
UN1282
azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717
C1=CC=NC=C1
79.102
CHEBI:16227
>99.9%
Pyridine, CHROMASOLV™ Plus, for HPLC, ≥99.9%
110-86-1
100.0000%
115°C
20°C
C5H5N
MFCD00011732
103233
JUJWROOIHBZHMG-UHFFFAOYSA-N
pyridine
1049
79.1
HPLC
Videos
Safety and Handling

Safety and Handling

ShelfLife : 450 days from date of manufacture

DOTInformation : Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Pyridine

EINECSNumber : 203-809-9

SDS
missing translation for 'documents'

missing translation for 'documents'