missing translation for 'onlineSavingsMsg'
Learn More

Pyridine, ≥99.9% (GC), MilliporeSigma™ Supelco™

Catalog No. 111020445
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020445 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020445 Supplier Merck Emd Millipore Supplier No. 024861ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 110-86-1
Ignition Point 899°F
Vapor Pressure 10 mmHg (13.2°C)
Vapor Density 2.72 (vs. air)
Density 0.978 g/mL (at 25°C)
Flash Point 17.0°C - closed cup
Linear Formula C5H5 N
MDL Number MFCD00011732
Melting Point -42°C
Boiling Point 115°C
Type Analytical Standard
Quantity 1 mL
Formula Weight 79.10
Percent Purity ≥99.9% (GC)
Grade Analytical Standard
Show More Show Less