missing translation for 'onlineSavingsMsg'
Learn More

Pyridine, 99.8%, for biochemistry, AcroSeal™, Thermo Scientific™

$356.40

Chemical Identifiers

CAS 110-86-1
Molecular Formula C5H5N
Molecular Weight (g/mol) 79.1
InChI Key JUJWROOIHBZHMG-UHFFFAOYSA-N
Synonym azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717
PubChem CID 1049
ChEBI CHEBI:16227
IUPAC Name pyridine
SMILES C1=CC=NC=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AC447900010
View Documents Promotion Details
N/A 1L N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

110-86-1
79.1
azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717
CHEBI:16227
C1=CC=NC=C1
C5H5N
JUJWROOIHBZHMG-UHFFFAOYSA-N
1049
pyridine

Specifications

110-86-1
115.0°C to 116.0°C
99.75% min. (GC)
C5H5N
0.978
azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717
JUJWROOIHBZHMG-UHFFFAOYSA-N
pyridine
1049
CHEBI:16227
99.8%
Pyridine
0.9780g/mL
17°C
AcroSeal™ Glass bottle
1.5085 to 1.5105
15,8081
Solubility in water: soluble. Other solubilities: soluble in alcohol, ether and most other common, organic solvents
C1=CC=NC=C1
79.1
0.95 mPa.s (20°C)
79.1
Biochemistry
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Highly flammable liquid and vapor.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

GHS Signal Word: Danger

EINECSNumber : 203-809-9

SDS
Documents

Documents

RUO – Research Use Only