Learn More
Pyridine-4-boronic acid hydrochloride, 95%
CAS: 913835-65-1 | C5H7BClNO2 | 159.38 g/mol
$84.01 - $315.99
Chemical Identifiers
| CAS | 913835-65-1 |
|---|---|
| Molecular Formula | C5H7BClNO2 |
| Molecular Weight (g/mol) | 159.38 |
| MDL Number | MFCD06201888 |
| InChI Key | NCZXYQJUECNKAD-UHFFFAOYSA-N |
| Synonym | pyridine-4-boronic acid hydrochloride, pyridin-4-ylboronic acid hydrochloride, pyridine-4-boronic acid, hcl, 4-pyridineboronic acid, hydrochloride, pyridine-4-boronic acid hcl, pyridine-4-boronic acid-hydrogen chloride, boronicacid, b-4-pyridinyl-, hydrochloride 1:1, c5h6bno2.hcl, pyridineboronic acid, 4-, hcl, 4-pyridineboronic acid, hcl |
| PubChem CID | 44118740 |
| IUPAC Name | pyridin-4-ylboronic acid;hydrochloride |
| SMILES | Cl.OB(O)C1=CC=NC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH5264803
|
Thermo Scientific Chemicals
H5264803 |
1 g |
Each for $84.01
|
|
|||||
|
AAH5264806
|
Thermo Scientific Chemicals
H5264806 |
5 g |
Each for $315.99
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 913835-65-1 | |
| 159.38 | |
| NCZXYQJUECNKAD-UHFFFAOYSA-N | |
| 44118740 | |
| Cl.OB(O)C1=CC=NC=C1 |
| C5H7BClNO2 | |
| MFCD06201888 | |
| pyridine-4-boronic acid hydrochloride, pyridin-4-ylboronic acid hydrochloride, pyridine-4-boronic acid, hcl, 4-pyridineboronic acid, hydrochloride, pyridine-4-boronic acid hcl, pyridine-4-boronic acid-hydrogen chloride, boronicacid, b-4-pyridinyl-, hydrochloride 1:1, c5h6bno2.hcl, pyridineboronic acid, 4-, hcl, 4-pyridineboronic acid, hcl | |
| pyridin-4-ylboronic acid;hydrochloride |
Specifications
| 913835-65-1 | |
| C5H7BClNO2 | |
| 1 g | |
| NCZXYQJUECNKAD-UHFFFAOYSA-N | |
| pyridin-4-ylboronic acid;hydrochloride | |
| 44118740 | |
| 95% |
| 340°C | |
| MFCD06201888 | |
| pyridine-4-boronic acid hydrochloride, pyridin-4-ylboronic acid hydrochloride, pyridine-4-boronic acid, hcl, 4-pyridineboronic acid, hydrochloride, pyridine-4-boronic acid hcl, pyridine-4-boronic acid-hydrogen chloride, boronicacid, b-4-pyridinyl-, hydrochloride 1:1, c5h6bno2.hcl, pyridineboronic acid, 4-, hcl, 4-pyridineboronic acid, hcl | |
| Cl.OB(O)C1=CC=NC=C1 | |
| 159.38 | |
| 159.38 | |
| Pyridine-4-boronic acid hydrochloride |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only