CAS RN 913835-65-1
IUPAC Name:
(pyridin-4-yl)boronic acid hydrochloride
Synonyms:
pyridin-4-ylboronic acid hydrochloride
Molecular Weight (g/mol):
159.38
Molecular Formula:
C5H7BClNO2
InChi Key:
NCZXYQJUECNKAD-UHFFFAOYSA-N
SMILES:
Cl.OB(O)C1=CC=NC=C1
1
–
1
of
1
results
1
Promotions Available
Pyridine-4-boronic acid hydrochloride, 95%
Pyridine-4-boronic acid hydrochloride, 95%
CAS: 913835-65-1 Molecular Formula: C5H7BClNO2 Molecular Weight (g/mol): 159.38 MDL Number: MFCD06201888 InChI Key: NCZXYQJUECNKAD-UHFFFAOYSA-N Synonym: pyridine-4-boronic acid hydrochloride,pyridin-4-ylboronic acid hydrochloride,pyridine-4-boronic acid, hcl,4-pyridineboronic acid, hydrochloride,pyridine-4-boronic acid hcl,pyridine-4-boronic acid-hydrogen chloride,boronicacid, b-4-pyridinyl-, hydrochloride 1:1,c5h6bno2.hcl,pyridineboronic acid, 4-, hcl,4-pyridineboronic acid, hcl PubChem CID: 44118740 IUPAC Name: pyridin-4-ylboronic acid;hydrochloride SMILES: Cl.OB(O)C1=CC=NC=C1