missing translation for 'onlineSavingsMsg'
Learn More

Pyridine-4-boronic acid hydrochloride, 95%

Catalog No. AAH5264803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5264803 1 g
AAH5264806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5264803 Supplier Thermo Scientific Chemicals Supplier No. H5264803
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 913835-65-1 | C5H7BClNO2 | 159.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 913835-65-1
Molecular Formula C5H7BClNO2
Molecular Weight (g/mol) 159.38
MDL Number MFCD06201888
InChI Key NCZXYQJUECNKAD-UHFFFAOYSA-N
Synonym pyridine-4-boronic acid hydrochloride, pyridin-4-ylboronic acid hydrochloride, pyridine-4-boronic acid, hcl, 4-pyridineboronic acid, hydrochloride, pyridine-4-boronic acid hcl, pyridine-4-boronic acid-hydrogen chloride, boronicacid, b-4-pyridinyl-, hydrochloride 1:1, c5h6bno2.hcl, pyridineboronic acid, 4-, hcl, 4-pyridineboronic acid, hcl
PubChem CID 44118740
IUPAC Name pyridin-4-ylboronic acid;hydrochloride
SMILES Cl.OB(O)C1=CC=NC=C1

Specifications

Melting Point 340°C
Quantity 1 g
Formula Weight 159.38
Percent Purity 95%
Chemical Name or Material Pyridine-4-boronic acid hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only