missing translation for 'onlineSavingsMsg'
Learn More

Pyridine-4-aldoxime 98.0+%, TCI America™

Catalog No. P067310G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P067310G 10 g
P067325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P067310G Supplier TCI America Supplier No. P067310G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 696-54-8
Molecular Formula C6H6N2O
Molecular Weight (g/mol) 122.13
MDL Number MFCD00006446
InChI Key OFYLBLSSPQTTHT-VMPITWQZSA-N
Synonym pyridine-4-aldoxime, 4-pyridinealdoxime, isonicotinaldehyde oxime, 4-pyridinecarboxaldehyde, oxime, isonicotinaldoxime, 4-hydroxyiminomethyl pyridine, 4-pyridinaldoxime, 4-pyridine aldoxime, pyridine, 4-formyl-, oxime, ccris 7744
PubChem CID 5324569
IUPAC Name (E)-N-[(pyridin-4-yl)methylidene]hydroxylamine
SMILES O\N=C\C1=CC=NC=C1

Specifications

Melting Point 133°C
Quantity 10 g
Formula Weight 122.13
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Pyridine-4-aldoxime
RTECSNumber NR9410000
TSCA No