accessibility menu, dialog, popup

UserName

OHNN(E)

CAS RN 696-54-8

IUPAC Name: (E)-N-[(pyridin-4-yl)methylidene]hydroxylamine
Synonyms: 4-pyridinealdoxime
Molecular Weight (g/mol): 122.13
Molecular Formula: C6H6N2O
InChi Key: OFYLBLSSPQTTHT-VMPITWQZSA-N
SMILES: O\N=C\C1=CC=NC=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)