missing translation for 'onlineSavingsMsg'
Learn More

Pyridine-3-boronic acid

CAS: 1692-25-7 | C5H6BNO2 | 122.92 g/mol

$134.52 - $134.52

Chemical Identifiers

CAS 1692-25-7
Molecular Formula C5H6BNO2
Molecular Weight (g/mol) 122.92
MDL Number MFCD00674177
InChI Key ABMYEXAYWZJVOV-UHFFFAOYSA-N
Synonym pyridine-3-boronic acid, 3-pyridylboronic acid, 3-pyridineboronic acid, pyridin-3-yl boronic acid, 3-pyridinylboronic acid, pyridin-3-yl-3-boronic acid, 3-pyridineboric acid, 3-pyridinebornic acid, pyridyl-3-boronic acid, pyridine 3-boronic acid
PubChem CID 2734378
IUPAC Name pyridin-3-ylboronic acid
SMILES OB(O)C1=CC=CN=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1504003
View Documents
Thermo Scientific Chemicals
L1504003
1 g
Each for $134.52
Only null left
 
Description

Description

Reactant used for preparation of phosphine-free Suzuki-Miyaura cross-coupling reactions, regioselective Suzuki-Miyaura coupling and tandem palladium-catalyzed intermolecular aminocarbonylation and annulation, N-arylation using copper acetylacetonate catalys, copper-mediated cyanation and regioselective cyanation of electron-rich benzenes, New linear poly(phenylpyridyl) chains by Suzuki coupling and Oligopyridyl foldamers as mimics of a-helix twist.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reactant used for preparation of phosphine-free Suzuki-Miyaura cross-coupling reactions, regioselective Suzuki-Miyaura coupling and tandem palladium-catalyzed intermolecular aminocarbonylation and annulation, N-arylation using copper acetylacetonate catalys, copper-mediated cyanation and regioselective cyanation of electron-rich benzenes, New linear poly(phenylpyridyl) chains by Suzuki coupling and Oligopyridyl foldamers as mimics of a-helix twist.

Solubility
Slightly soluble in methanol.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Chemical Identifiers

1692-25-7
122.92
ABMYEXAYWZJVOV-UHFFFAOYSA-N
2734378
OB(O)C1=CC=CN=C1
C5H6BNO2
MFCD00674177
pyridine-3-boronic acid, 3-pyridylboronic acid, 3-pyridineboronic acid, pyridin-3-yl boronic acid, 3-pyridinylboronic acid, pyridin-3-yl-3-boronic acid, 3-pyridineboric acid, 3-pyridinebornic acid, pyridyl-3-boronic acid, pyridine 3-boronic acid
pyridin-3-ylboronic acid

Specifications

1692-25-7
C5H6BNO2
1 g
pyridine-3-boronic acid, 3-pyridylboronic acid, 3-pyridineboronic acid, pyridin-3-yl boronic acid, 3-pyridinylboronic acid, pyridin-3-yl-3-boronic acid, 3-pyridineboric acid, 3-pyridinebornic acid, pyridyl-3-boronic acid, pyridine 3-boronic acid
ABMYEXAYWZJVOV-UHFFFAOYSA-N
pyridin-3-ylboronic acid
2734378
Pyridine-3-boronic acid
>300°C
MFCD00674177
471943
Slightly soluble in methanol.
OB(O)C1=CC=CN=C1
122.92
122.92
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335-H500

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only