Learn More
Pyridine-3-boronic acid
CAS: 1692-25-7 | C5H6BNO2 | 122.92 g/mol
Supplier: Thermo Scientific Chemicals L1504003
Description
Reactant used for preparation of phosphine-free Suzuki-Miyaura cross-coupling reactions, regioselective Suzuki-Miyaura coupling and tandem palladium-catalyzed intermolecular aminocarbonylation and annulation, N-arylation using copper acetylacetonate catalys, copper-mediated cyanation and regioselective cyanation of electron-rich benzenes, New linear poly(phenylpyridyl) chains by Suzuki coupling and Oligopyridyl foldamers as mimics of a-helix twist.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsReactant used for preparation of phosphine-free Suzuki-Miyaura cross-coupling reactions, regioselective Suzuki-Miyaura coupling and tandem palladium-catalyzed intermolecular aminocarbonylation and annulation, N-arylation using copper acetylacetonate catalys, copper-mediated cyanation and regioselective cyanation of electron-rich benzenes, New linear poly(phenylpyridyl) chains by Suzuki coupling and Oligopyridyl foldamers as mimics of a-helix twist.
Solubility
Slightly soluble in methanol.
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
| Pyridine-3-boronic acid | |
| >300°C | |
| MFCD00674177 | |
| 471943 | |
| Slightly soluble in methanol. | |
| OB(O)C1=CC=CN=C1 | |
| 122.92 | |
| 122.92 |
| 1692-25-7 | |
| C5H6BNO2 | |
| 1 g | |
| pyridine-3-boronic acid, 3-pyridylboronic acid, 3-pyridineboronic acid, pyridin-3-yl boronic acid, 3-pyridinylboronic acid, pyridin-3-yl-3-boronic acid, 3-pyridineboric acid, 3-pyridinebornic acid, pyridyl-3-boronic acid, pyridine 3-boronic acid | |
| ABMYEXAYWZJVOV-UHFFFAOYSA-N | |
| pyridin-3-ylboronic acid | |
| 2734378 |
Chemical Identifiers
| 1692-25-7 | |
| 122.92 | |
| ABMYEXAYWZJVOV-UHFFFAOYSA-N | |
| 2734378 | |
| OB(O)C1=CC=CN=C1 |
| C5H6BNO2 | |
| MFCD00674177 | |
| pyridine-3-boronic acid, 3-pyridylboronic acid, 3-pyridineboronic acid, pyridin-3-yl boronic acid, 3-pyridinylboronic acid, pyridin-3-yl-3-boronic acid, 3-pyridineboric acid, 3-pyridinebornic acid, pyridyl-3-boronic acid, pyridine 3-boronic acid | |
| pyridin-3-ylboronic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335-H500
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only