missing translation for 'onlineSavingsMsg'
Learn More

Pyridine, 25% v/v, Reagents

Catalog No. 01334425
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01334425 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01334425 Supplier Labchem Inc Supplier No. CP1040004C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pyridine, 25% v/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 110-86-1
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Azabenzene, Azine
IUPAC Name water
SMILES O

Specifications

Packaging Glass Bottle
Quantity 4 L
Specific Gravity 1.05
Grade Reagent
Physical Form Liquid
Chemical Name or Material Pyridine, 25% v/v