missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyridine-2-sulfonic Acid 98.0+%, TCI America™
Supplier: TCI America P164725G
Specifications
| Pyridine-2-sulfonic Acid | |
| 252°C | |
| C5H5NO3S | |
| 25 g | |
| 2-pyridinesulfonic acid, pyridinesulfonic acid, 2-pyridinesulfonic acid 8ci 9ci, alpha-pyridinesulfonic acid, sulfopyridin, pyridine-2-sulfonicacid, acmc-1bo1h, .alpha.-pyridinesulfonic acid, kzvlnagysakymg-uhfffaoysa | |
| OS(=O)(=O)C1=CC=CC=N1 | |
| 159.16 | |
| 159.16 | |
| Crystalline Powder |
| 15103-48-7 | |
| White-Yellow | |
| MFCD00234080 | |
| 3261 | |
| KZVLNAGYSAKYMG-UHFFFAOYSA-N | |
| pyridine-2-sulfonic acid | |
| 94849 | |
| ≥98.0% (HPLC,T) |
Chemical Identifiers
| 15103-48-7 | |
| 159.16 | |
| KZVLNAGYSAKYMG-UHFFFAOYSA-N | |
| 94849 | |
| OS(=O)(=O)C1=CC=CC=N1 |
| C5H5NO3S | |
| MFCD00234080 | |
| 2-pyridinesulfonic acid, pyridinesulfonic acid, 2-pyridinesulfonic acid 8ci 9ci, alpha-pyridinesulfonic acid, sulfopyridin, pyridine-2-sulfonicacid, acmc-1bo1h, .alpha.-pyridinesulfonic acid, kzvlnagysakymg-uhfffaoysa | |
| pyridine-2-sulfonic acid |
Safety and Handling
TSCA : No