missing translation for 'onlineSavingsMsg'
Learn More

Pyridine-2-sulfonic Acid 98.0+%, TCI America™

Catalog No. p-7135191
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P16475G 5 g
P164725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P16475G Supplier TCI America Supplier No. P16475G

Chemical Identifiers

CAS 15103-48-7
Molecular Formula C5H5NO3S
Molecular Weight (g/mol) 159.16
MDL Number MFCD00234080
InChI Key KZVLNAGYSAKYMG-UHFFFAOYSA-N
Synonym 2-pyridinesulfonic acid, pyridinesulfonic acid, 2-pyridinesulfonic acid 8ci 9ci, alpha-pyridinesulfonic acid, sulfopyridin, pyridine-2-sulfonicacid, acmc-1bo1h, .alpha.-pyridinesulfonic acid, kzvlnagysakymg-uhfffaoysa
PubChem CID 94849
IUPAC Name pyridine-2-sulfonic acid
SMILES OS(=O)(=O)C1=CC=CC=N1

Specifications

Melting Point 252°C
Color White-Yellow
Quantity 5 g
UN Number 3261
Formula Weight 159.16
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Pyridine-2-sulfonic Acid