missing translation for 'onlineSavingsMsg'
Learn More

Pyridine-2-carboxylic Acid 99.0+%, TCI America™

Catalog No. p-7134598
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Catalog No. Quantity
P042125G 25 g
P0421100G 100 g
P0421500G 500 g
3 options

Catalog No. P042125G

Supplier: TCI America P042125G

Only null left

Chemical Identifiers

CAS 98-98-6
Molecular Formula C6H5NO2
Molecular Weight (g/mol) 123.111
MDL Number MFCD00006293
InChI Key SIOXPEMLGUPBBT-UHFFFAOYSA-N
Synonym picolinic acid, 2-pyridinecarboxylic acid, 2-picolinic acid, 2-carboxypyridine, alpha-picolinic acid, o-pyridinecarboxylic acid, pyridinecarboxylic acid, alpha-pyridinecarboxylic acid, picolinate, acide picolique
PubChem CID 1018
ChEBI CHEBI:28747
IUPAC Name pyridine-2-carboxylic acid
SMILES C1=CC=NC(=C1)C(=O)O

Specifications

Melting Point 140°C
Color White
Quantity 25 g
Formula Weight 123.11
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Pyridine-2-carboxylic Acid
EINECSNumber (9)-1077
RTECSNumber TJ7344000
TSCA Yes