accessibility menu, dialog, popup

UserName

CAS RN 98-98-6

IUPAC Name: pyridine-2-carboxylic acid
Synonyms: picolinic acid acide picolique α-picolinic acid L-baikiain 2-picolinic acid pyridine-carboxylique-2
Molecular Weight (g/mol): 123.11
Molecular Formula: C6H5NO2
InChi Key: SIOXPEMLGUPBBT-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC=N1
Molecular Weight (g/mol) 
  • (4)
  • (6)
Percent Purity 
  • (3)
  • (1)
  • (6)
Quantity 
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
Physical Form 
  • (7)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (7)
  • (4)
  • (6)
  • (3)
  • (1)
  • (6)
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (7)
  • (1)