missing translation for 'onlineSavingsMsg'
Learn More

Pyridine-2-carboxaldoxime methochloride, 97%

Catalog No. AAB2389006
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2389006 5 g
AAB2389014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2389006 Supplier Thermo Scientific Chemicals Supplier No. B2389006

CAS: 51-15-0 | C7H9ClN2O | 172.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 51-15-0
Molecular Formula C7H9ClN2O
Molecular Weight (g/mol) 172.61
MDL Number MFCD00011981
InChI Key HIGSLXSBYYMVKI-UHDJGPCESA-N
Synonym pralidoxime chloride, 1-methylpyridinium-2-aldoxime chloride, 2-pam-cl, 2-hydroxyimino methyl-1-methylpyridin-1-ium chloride, 2-hydroxyimino methyl-1-methylpyridinium chloride, 2-formyl 1-methyl pyridinium oxime chloride
PubChem CID 57428996
SMILES Cl.CN1C=CC=C\C1=C/N=O

Specifications

Melting Point ∼223°C (decomposition)
Quantity 5 g
Beilstein 4163981
Merck Index 14,7703
IUPAC Name [(Z)-(1-methylpyridin-2-ylidene)methyl]-oxoazanium;chloride
Formula Weight 172.62
Percent Purity 97%
Chemical Name or Material Pyridine-2-carboxaldoxime methochloride
Hazard Category H302+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 200-080-9
RTECSNumber UU4200000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only