missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyridine-2-carbonyl Chloride Hydrochloride 93.0+%, TCI America™
Supplier: TCI America P061925G
Specifications
| Pyridine-2-carbonyl Chloride Hydrochloride | |
| Red-White | |
| MFCD00060193 | |
| 3261 | |
| VIPHVHVAGBKHGR-UHFFFAOYSA-N | |
| hydrogen pyridine-2-carbonyl chloride chloride | |
| 135033 | |
| ≥93.0% (T) |
| 39901-94-5 | |
| C6H5Cl2NO | |
| 25 g | |
| Picolinoyl Chloride Hydrochloride | |
| [H+].[Cl-].ClC(=O)C1=CC=CC=N1 | |
| 178.01 | |
| 178.01 | |
| Crystalline Powder |
Chemical Identifiers
| 39901-94-5 | |
| 178.01 | |
| VIPHVHVAGBKHGR-UHFFFAOYSA-N | |
| 135033 | |
| [H+].[Cl-].ClC(=O)C1=CC=CC=N1 |
| C6H5Cl2NO | |
| MFCD00060193 | |
| Picolinoyl Chloride Hydrochloride | |
| hydrogen pyridine-2-carbonyl chloride chloride |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator