missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyridine-2-carbonyl Chloride Hydrochloride 93.0+%, TCI America™
$130.48 - $331.81
Chemical Identifiers
| CAS | 39901-94-5 |
|---|---|
| Molecular Formula | C6H5Cl2NO |
| Molecular Weight (g/mol) | 178.01 |
| MDL Number | MFCD00060193 |
| InChI Key | VIPHVHVAGBKHGR-UHFFFAOYSA-N |
| Synonym | Picolinoyl Chloride Hydrochloride |
| PubChem CID | 135033 |
| IUPAC Name | hydrogen pyridine-2-carbonyl chloride chloride |
| SMILES | [H+].[Cl-].ClC(=O)C1=CC=CC=N1 |
Chemical Identifiers
| 39901-94-5 | |
| 178.01 | |
| VIPHVHVAGBKHGR-UHFFFAOYSA-N | |
| 135033 | |
| [H+].[Cl-].ClC(=O)C1=CC=CC=N1 |
| C6H5Cl2NO | |
| MFCD00060193 | |
| Picolinoyl Chloride Hydrochloride | |
| hydrogen pyridine-2-carbonyl chloride chloride |
Specifications
| 39901-94-5 | |
| C6H5Cl2NO | |
| 5 g | |
| Picolinoyl Chloride Hydrochloride | |
| [H+].[Cl-].ClC(=O)C1=CC=CC=N1 | |
| 178.01 | |
| 178.01 | |
| Crystalline Powder |
| Red-White | |
| MFCD00060193 | |
| 3261 | |
| VIPHVHVAGBKHGR-UHFFFAOYSA-N | |
| hydrogen pyridine-2-carbonyl chloride chloride | |
| 135033 | |
| ≥93.0% (T) | |
| Pyridine-2-carbonyl Chloride Hydrochloride |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator