Learn More
Pyridine-2,6-dicarbonyl dichloride, 97%
CAS: 3739-94-4 | C7H3Cl2NO2 | 204.006 g/mol
Supplier: Thermo Scientific Chemicals H2657006
Description
2,6-Pyridinedicarbonyl dichloride is used as the reagent during the synthesis of pyridine-based polyamido-polyester optically active macrocycles. It is used as starting reagent during the synthesis of pyridine-bridged 2,6-bis-carboxamide Schiff?s bases. It was used in the synthesis of N,N?-bis(1-pyrenylmethyl)pyridine-2,6-dicarboxamide3
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,6-Pyridinedicarbonyl dichloride is used as the reagent during the synthesis of pyridine-based polyamido-polyester optically active macrocycles. It is used as starting reagent during the synthesis of pyridine-bridged 2,6-bis-carboxamide Schiff′s bases. It was used in the synthesis of N,N′-bis(1-pyrenylmethyl)pyridine-2,6-dicarboxamide3
Solubility
Insoluble in water.
Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
| Pyridine-2,6-dicarbonyl dichloride | |
| 57°C to 61°C | |
| Irritating | |
| MFCD00006289 | |
| UN3261 | |
| Moisture sensitive | |
| Insoluble in water. | |
| C1=CC(=NC(=C1)C(=O)Cl)C(=O)Cl | |
| 204.006 | |
| 204.01 |
| 3739-94-4 | |
| 284°C | |
| C7H3Cl2NO2 | |
| 5 g | |
| 131556 | |
| 2,6-pyridinedicarbonyl dichloride, 2,6-pyridinedicarboxylic acid chloride, pyridine-2,6-dicarbonyl dichloride, 2,6-pyridinedicarbonyl chloride, pyridine-2,6-dicarboxylic acid chloride, 2,6-pyridinedicarboxylicacidchloride, pyridine-2,6-dicarboyl dichloride, 2,6-pyridinedicarbony chloride, pubchem13243, tpc-i153 | |
| GWHOGODUVLQCEB-UHFFFAOYSA-N | |
| pyridine-2,6-dicarbonyl chloride | |
| 77335 | |
| 98% |
Chemical Identifiers
| 3739-94-4 | |
| 204.006 | |
| GWHOGODUVLQCEB-UHFFFAOYSA-N | |
| 77335 | |
| C1=CC(=NC(=C1)C(=O)Cl)C(=O)Cl |
| C7H3Cl2NO2 | |
| MFCD00006289 | |
| 2,6-pyridinedicarbonyl dichloride, 2,6-pyridinedicarboxylic acid chloride, pyridine-2,6-dicarbonyl dichloride, 2,6-pyridinedicarbonyl chloride, pyridine-2,6-dicarboxylic acid chloride, 2,6-pyridinedicarboxylicacidchloride, pyridine-2,6-dicarboyl dichloride, 2,6-pyridinedicarbony chloride, pubchem13243, tpc-i153 | |
| pyridine-2,6-dicarbonyl chloride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 223-125-4
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only