missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyridine-2,3-dicarbonitrile 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 79°C |
| Color | White |
| Quantity | 5 g |
| UN Number | 3439 |
| Formula Weight | 129.12 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Pyridine-2,3-dicarbonitrile |
Safety and Handling
| TSCA | No |