accessibility menu, dialog, popup

UserName

CAS RN 17132-78-4

NNN

CAS RN 17132-78-4

IUPAC Name: pyridine-2,3-dicarbonitrile
Synonyms: pyridine-2,3-dicarbonitrile
Molecular Weight (g/mol): 129.12
Molecular Formula: C7H3N3
InChi Key: GHFGOVUYCKZOJH-UHFFFAOYSA-N
SMILES: N#CC1=CC=CN=C1C#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results