missing translation for 'onlineSavingsMsg'
Learn More

Pyridine-2,3-dicarbonitrile 98.0+%, TCI America™

Catalog No. P12181G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P12181G 1 g
P12185G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P12181G Supplier TCI America Supplier No. P12181G
Only null left

Chemical Identifiers

CAS 17132-78-4
Molecular Formula C7H3N3
Molecular Weight (g/mol) 129.12
MDL Number MFCD00191682
InChI Key GHFGOVUYCKZOJH-UHFFFAOYSA-N
Synonym 2,3-Dicyanopyridine
PubChem CID 28302
IUPAC Name pyridine-2,3-dicarbonitrile
SMILES N#CC1=CC=CN=C1C#N

Specifications

Melting Point 79°C
Color White
Quantity 1 g
UN Number 3439
Formula Weight 129.12
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Pyridine-2,3-dicarbonitrile