missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyridazine 99.0+%, TCI America™
$109.98 - $324.35
Chemical Identifiers
| CAS | 289-80-5 |
|---|---|
| Molecular Formula | C4H4N2 |
| Molecular Weight (g/mol) | 80.09 |
| MDL Number | MFCD00006463 |
| InChI Key | PBMFSQRYOILNGV-UHFFFAOYSA-N |
| Synonym | orthodiazine, 1,2-diazine, o-diazine, 1,2-diazabenzene, pyridazin, 1,2-diazin, oizine, pyridazine, pubchem17789, acmc-209h5x |
| PubChem CID | 9259 |
| ChEBI | CHEBI:30954 |
| IUPAC Name | pyridazine |
| SMILES | C1=CC=NN=C1 |
Chemical Identifiers
| 289-80-5 | |
| 80.09 | |
| PBMFSQRYOILNGV-UHFFFAOYSA-N | |
| 9259 | |
| pyridazine |
| C4H4N2 | |
| MFCD00006463 | |
| orthodiazine, 1,2-diazine, o-diazine, 1,2-diazabenzene, pyridazin, 1,2-diazin, oizine, pyridazine, pubchem17789, acmc-209h5x | |
| CHEBI:30954 | |
| C1=CC=NN=C1 |
Specifications
| 289-80-5 | |
| Yellow | |
| C4H4N2 | |
| 5 g | |
| PBMFSQRYOILNGV-UHFFFAOYSA-N | |
| pyridazine | |
| 9259 | |
| 80.09 | |
| Liquid |
| -8°C | |
| 208°C | |
| MFCD00006463 | |
| orthodiazine, 1,2-diazine, o-diazine, 1,2-diazabenzene, pyridazin, 1,2-diazin, oizine, pyridazine, pubchem17789, acmc-209h5x | |
| C1=CC=NN=C1 | |
| 80.09 | |
| CHEBI:30954 | |
| ≥99.0% (GC) | |
| Pyridazine |
Safety and Handling
RTECSNumber : UR5267000
TSCA : Yes