accessibility menu, dialog, popup

UserName

NN

CAS RN 289-80-5

IUPAC Name: pyridazine
Synonyms: pyridazine
Molecular Weight (g/mol): 80.09
Molecular Formula: C4H4N2
InChi Key: PBMFSQRYOILNGV-UHFFFAOYSA-N
SMILES: C1=CC=NN=C1
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)