missing translation for 'onlineSavingsMsg'
Learn More

Pyridazine 99.0+%, TCI America™

Catalog No. P06165G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P06165G 5 g
P061625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P06165G Supplier TCI America Supplier No. P06165G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 289-80-5
Molecular Formula C4H4N2
Molecular Weight (g/mol) 80.09
MDL Number MFCD00006463
InChI Key PBMFSQRYOILNGV-UHFFFAOYSA-N
Synonym orthodiazine, 1,2-diazine, o-diazine, 1,2-diazabenzene, pyridazin, 1,2-diazin, oizine, pyridazine, pubchem17789, acmc-209h5x
PubChem CID 9259
ChEBI CHEBI:30954
IUPAC Name pyridazine
SMILES C1=CC=NN=C1

Specifications

Melting Point -8°C
Color Yellow
Boiling Point 208°C
Quantity 5 g
Formula Weight 80.09
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Pyridazine
RTECSNumber UR5267000
TSCA Yes