missing translation for 'onlineSavingsMsg'
Learn More

Pyridazine, 98+%

Catalog No. AAA1326606
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1326606 5 g
AAA1326614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1326606 Supplier Thermo Scientific Chemicals Supplier No. A1326606

CAS: 289-80-5 | C4H4N2 | 80.09 g/mol

Pyrrolo[1,2-b]pyridazine 1 is a N-bridgehead aromatic heterocycle containing two nitrogen atoms, is obtained by the condensation of pyridazine and pyrrole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyrrolo[1,2-b]pyridazine 1 is a N-bridgehead aromatic heterocycle containing two nitrogen atoms, is obtained by the condensation of pyridazine and pyrrole.

Solubility
Miscible with water, benzene, DMF. Soluble in alcohol, ether

Notes
Store at room temperature. Incompatible with oxidizing agents. Air sensitive.

Chemical Identifiers

CAS 289-80-5
Molecular Formula C4H4N2
Molecular Weight (g/mol) 80.09
MDL Number MFCD00006463
InChI Key PBMFSQRYOILNGV-UHFFFAOYSA-N
Synonym orthodiazine, 1,2-diazine, o-diazine, 1,2-diazabenzene, pyridazin, 1,2-diazin, oizine, pyridazine, pubchem17789, acmc-209h5x
PubChem CID 9259
ChEBI CHEBI:30954
IUPAC Name pyridazine
SMILES C1=CC=NN=C1

Specifications

Melting Point -8°C
Density 1.105
Boiling Point 87°C (14 mmHg)
Flash Point 85°C (185°F)
Odor Unpleasant
Refractive Index 1.524
Quantity 5 g
Beilstein 103906
Merck Index 14,7969
Solubility Information Miscible with water, benzene, DMF; Soluble in alcohol, ether
Formula Weight 80.09
Percent Purity ≥98%
Chemical Name or Material Pyridazine
Show More Show Less
Hazard Category H227
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 206-025-5
RTECSNumber UR5267000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only