missing translation for 'onlineSavingsMsg'
Learn More

Corning™ PYREX™ Ribbed Watch Glass/Beaker Cover

Catalog No. 02613C
Change view
Click to view available options
Diameter (Metric):
7.5 cm
10 cm
12.5 cm
For Use With (Equipment):
For 1000 mL beaker
For 250 mL beaker
For 600 mL beaker
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) For Use With (Equipment)
02613C 12.5 cm For 1000 mL beaker
02613A 7.5 cm For 250 mL beaker
02613B 10 cm For 600 mL beaker
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 02613C Supplier Corning™ Supplier No. 9990125
Get Up to $2,500 in Products with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ribbed design permits reflux of digestate and escape of excess vapor, thus preventing vapor buildup.

Designed for use as a beaker cover. Ribs are molded into the glass so that they can’t pop off as they do when cane is sealed to standard watch glasses. Permits reflux of digestate and escape of excess vapor. Eliminates risk of contamination from dropped hooks or props. Meets EPA requirements for methods 3005, 3010, and 3020.

Compliance

Meet U.S. EPA requirements for Methods 3005, 3010, and 3020.

Specifications

Certifications/Compliance U.S. EPA
Disposable Reusable
Material Borosilicate Glass
Shape Round
Type Watch Glass
For Use With (Application) Used as Beaker Covers
For Use With (Equipment) For 1000 mL beaker
Interior Finish Rib
No. per Case 12/Cs.
No. per Pack 12/Pk.
Diameter (English) 4.92 in.
Diameter (Metric) 12.5 cm
Show More Show Less