Learn More
Corning™ PYREX™ Replacement Friedrichs Condenser for 250 mL Extractor System
GREENER_CHOICE
Catalog No. 0957110B
Change view
Click to view available options
Standard Taper Joint Bottom:
45/50
Catalog No. Standard Taper Joint Bottom
0957110B 45/50
1 options
Catalog No. 0957110B Supplier Corning™ Supplier No. 3880MCO
Get Up to $2,500 in Products with Purchase
View all promotions
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Condenser only
This is a replacement PYREX™ Friedrichs condenser for the 250 mL Extractor System. This special Friedrichs condenser has a 45/50 inner Standard Taper joint on the bottom. The condenser tubulation O.D. is 10mm.
Specifications
Product Type Friedrichs Condenser
For Use With (Equipment) Extractor System 250 mL
Standard Taper Joint Bottom 45/50